Learn More
Medchemexpress LLC Pomalidomide-PEG4-C2-NH2 hydrochloride | 2357105-92-9 | 98.2% | 528.98 | 500 MG
SDP
Catalog No. 5000436233
Encompass_Preferred
Pomalidomide-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate used in the synthesis of PROTACs. It incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Used with a 4-unit PEG linker
  • Applicable in the synthesis of PROTACs
  • Intended for research use only
  • Not for patient use
Catalog No. 50-004-36233 Supplier Medchemexpress LLC Supplier No. HY112599B500MG
Pomalidomide-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate used in the synthesis of PROTACs. It incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Used with a 4-unit PEG linker
  • Applicable in the synthesis of PROTACs
  • Intended for research use only
  • Not for patient use