Learn More
Medchemexpress LLC Pomalidomide-PEG4-COOH | 2138440-81-8 | 98.0% | 521.52 | 250 MG
SDP
Catalog No. 5000366221
Encompass_Preferred
Pomalidomide-PEG4-COOH is an E3 ligase ligand-linker conjugate used in PROTAC technology. It contains a Pomalidomide-based cereblon ligand and a 4-unit PEG linker.
  • E3 ligase ligand-linker conjugate
  • Contains Pomalidomide-based cereblon ligand
  • Features a 4-unit PEG linker
  • Used in PROTAC technology
Catalog No. 50-003-66221 Supplier Medchemexpress LLC Supplier No. 250MGHY133699
Pomalidomide-PEG4-COOH is an E3 ligase ligand-linker conjugate used in PROTAC technology. It contains a Pomalidomide-based cereblon ligand and a 4-unit PEG linker.
  • E3 ligase ligand-linker conjugate
  • Contains Pomalidomide-based cereblon ligand
  • Features a 4-unit PEG linker
  • Used in PROTAC technology