Learn More
Medchemexpress LLC Pomalidomide-PEG6-C2-COOH | 2225148-49-0 | MFCD32173791 | >98.0% | C28H39N3O12 | 10MG
SDP
Catalog No. 5000259454
Encompass_Preferred
Pomalidomide-PEG6-C2-COOH is a cereblon E3 ligase ligand-linker conjugate that combines a pomalidomide-derived cereblon binder with a six-unit polyethylene glycol spacer terminating in a carboxylic acid. Supplied as a high-purity building block, it is intended for use in the synthesis of PROTACs and other targeted protein degradation molecules, enabling modular attachment to warheads or additional linker chemistry.
  • Cereblon-binding pomalidomide core for E3 ligase recruitment.
  • PEG6 spacer provides a flexible, water-compatible linker.
  • Terminal carboxylic acid enables amide coupling and conjugation.
  • High chemical purity suitable for medicinal chemistry research.
  • Available in multiple small-scale package sizes for screening and optimisation.
  • Stable under recommended storage conditions for reliable handling.
Catalog No. 50-002-59454 Supplier Medchemexpress LLC Supplier No. HY14165810MG
Pomalidomide-PEG6-C2-COOH is a cereblon E3 ligase ligand-linker conjugate that combines a pomalidomide-derived cereblon binder with a six-unit polyethylene glycol spacer terminating in a carboxylic acid. Supplied as a high-purity building block, it is intended for use in the synthesis of PROTACs and other targeted protein degradation molecules, enabling modular attachment to warheads or additional linker chemistry.
  • Cereblon-binding pomalidomide core for E3 ligase recruitment.
  • PEG6 spacer provides a flexible, water-compatible linker.
  • Terminal carboxylic acid enables amide coupling and conjugation.
  • High chemical purity suitable for medicinal chemistry research.
  • Available in multiple small-scale package sizes for screening and optimisation.
  • Stable under recommended storage conditions for reliable handling.