Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-PEG6-C2-COOH | 2225148-49-0 | MFCD32173791 | 99.0% | C28H39N3O12 | 50 MG
SDP

Catalog No. 5000353337
Encompass_Preferred

Pomalidomide-PEG6-C2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 6-unit PEG linker used in PROTAC technology. This product is for research use only.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Features a 6-unit PEG linker
  • Used in PROTAC technology
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Shipping: Room temperature in continental US; may vary elsewhere

Catalog No. 50-003-53337 Supplier Medchemexpress LLC Supplier No. HY14165850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-PEG6-C2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 6-unit PEG linker used in PROTAC technology. This product is for research use only.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Features a 6-unit PEG linker
  • Used in PROTAC technology
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Shipping: Room temperature in continental US; may vary elsewhere

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.