Learn More
Medchemexpress LLC Pomalidomide-PEG6-NH2 (hydrochloride) | 2341841-01-6 | 98.01% | 574.02 | 25 MG
SDP
Catalog No. 5000350188
Encompass_Preferred
Pomalidomide-PEG6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide-based cereblon ligand and a 6-unit PEG linker, used in PROTAC technology.
  • Incorporates pomalidomide-based cereblon ligand
  • Features 6-unit PEG linker
  • Used in PROTAC technology
  • Targets cereblon
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50188 Supplier Medchemexpress LLC Supplier No. 25MGHY136161
Pomalidomide-PEG6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide-based cereblon ligand and a 6-unit PEG linker, used in PROTAC technology.
  • Incorporates pomalidomide-based cereblon ligand
  • Features 6-unit PEG linker
  • Used in PROTAC technology
  • Targets cereblon
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins