Learn More
Medchemexpress LLC Pomalidomide-PEG6-NH2 hydrochloride | 2341841-01-6 | MFCD32734576 | 98.0% | 574.02 g/mol | C25H36ClN3O10 | 5 MG
SDP
Catalog No. 5000221898
Encompass_Preferred
Pomalidomide-PEG6-NH2 hydrochloride is a pomalidomide-based cereblon (CRBN) E3 ligase ligand conjugated to a six-unit polyethylene glycol (PEG6) linker. Supplied as the hydrochloride salt, it is intended as a ligand-linker building block for PROTAC design and targeted protein degradation research.
  • Pomalidomide-based CRBN recruiter for use in PROTAC assembly.
  • Six-unit PEG linker provides flexible spacing between ligand and warhead.
  • Hydrochloride salt form supports handling and solubility in common solvents.
  • High purity suitable for medicinal chemistry (98.0% by LCMS reported).
  • Available in milligram pack sizes appropriate for synthesis and assay development.
Catalog No. 50-002-21898 Supplier Medchemexpress LLC Supplier No. HY1361615MG
Pomalidomide-PEG6-NH2 hydrochloride is a pomalidomide-based cereblon (CRBN) E3 ligase ligand conjugated to a six-unit polyethylene glycol (PEG6) linker. Supplied as the hydrochloride salt, it is intended as a ligand-linker building block for PROTAC design and targeted protein degradation research.
  • Pomalidomide-based CRBN recruiter for use in PROTAC assembly.
  • Six-unit PEG linker provides flexible spacing between ligand and warhead.
  • Hydrochloride salt form supports handling and solubility in common solvents.
  • High purity suitable for medicinal chemistry (98.0% by LCMS reported).
  • Available in milligram pack sizes appropriate for synthesis and assay development.