Learn More
Aviva Systems Biology Corp Pomgnt1 Antibody - N-terminal region (ARP48808_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pomgnt1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 0610016I07Rik, 4930467B06Rik, RP23-319H15.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-380