Learn More
Medchemexpress LLC HY-131904 5mg , POMHEX CAS:2004714-34-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1319045MG
Encompass_Preferred
HY-131904 5mg POMHEX CAS: 2004714-34-3 POMHEX, a racemic mixture and a cell-permeable pivaloyloxymethyl (POM) prodrug of HEX, is a potent, ENO2-specific inhibitor of enolase. POMHEX exhibits low-nanomolar potency against ENO1-deleted cells in vitro and is capable of eradicating ENO1-deleted xenografted tumours in vivo. POMHEX is a potent glycolysis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9240