Learn More
Enzo Life Sciences POMP polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW97150100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 129-141 of human POMP. Species reactivity: Human, Rat, Monkey. Applications: IHC, IP, WB. Purity: Purified by salt precipitation and peptide immunochromatography. Formulation: Liquid.  In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1700