Learn More
Apexbio Technology LLC β-Pompilidotoxin 216064-36-7 10mg
SDP
Catalog No. 5000095574
Encompass_Preferred
-Pompilidotoxin ( -PMTX CAS 216064-36-7) is a peptide neurotoxin isolated from the venom of the solitary wasp Batozonellus maculifrons Composed of 13 amino acids (C71H124N22O17) -PMTX modulates neuronal excitability by selectively slowing the inactivation of voltage-gated sodium (Na ) channels without affecting cardiac Na channels This action is thought to involve interaction with neurotoxin receptor site 3 on the extracellular surface of neuronal Na channels Compared to -pompilidotoxin -PMTX exhibits greater potency in enhancing synaptic transmission at neuromuscular junctions -PMTX serves as a valuable tool for investigating Na channel kinetics and the molecular mechanisms regulating synaptic transmission
Catalog No. 50-000-95574 Supplier Apexbio Technology LLC Supplier No. A113210
-Pompilidotoxin ( -PMTX CAS 216064-36-7) is a peptide neurotoxin isolated from the venom of the solitary wasp Batozonellus maculifrons Composed of 13 amino acids (C71H124N22O17) -PMTX modulates neuronal excitability by selectively slowing the inactivation of voltage-gated sodium (Na ) channels without affecting cardiac Na channels This action is thought to involve interaction with neurotoxin receptor site 3 on the extracellular surface of neuronal Na channels Compared to -pompilidotoxin -PMTX exhibits greater potency in enhancing synaptic transmission at neuromuscular junctions -PMTX serves as a valuable tool for investigating Na channel kinetics and the molecular mechanisms regulating synaptic transmission