Learn More
Aviva Systems Biology Corp POMT1 Antibody - middle region (ARP44994_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44994_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POMT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human POMT1. Alternate names: FLJ37239, LGMD2K, RT, MDDGA1, MDDGB1, MDDGC1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58950