Learn More
Aviva Systems Biology Corp PON1 Antibody - C-terminal region (ARP41401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PON1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PON1. Alternate names: ESA, PON, MVCD5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57833