Learn More
Aviva Systems Biology Corp PON3 Antibody - middle region (ARP41832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PON3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PON3. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58010