Learn More
Cayman Chemical Ponasterone A, Purity: ≥95%, CAS: 13408-56-5, 10 mg
SDP
Catalog No. 502970173
Encompass_Preferred
An insect hormone used as an inducer of ecdysone-inducible mammalian expression systems
Catalog No. 50-297-0173 Supplier Cayman Chemical Supplier No. 1638610 MG
An insect hormone used as an inducer of ecdysone-inducible mammalian expression systems