Learn More
Enzo Life Sciences Ponasterone A (5mg). CAS: 13408-56-5
SDP
Supplier:  Enzo Life Sciences ALX370014M005
Encompass_Preferred
A member of the ecdysteroid family. Analog of ecdysone with similar properties to muristerone A. It is a functional, reliable and economical substitute for muristerone A as an inducer for the ecdysone-inducible mammalian expression system. Results show that ponasterone A was able to induce expression of β-galactosidase to levels similar to those obtained with muristerone A induction. Potent compound for ecdysteroid activity; EC50= 0.07 µM. Alternative name: 25-Deoxy-20-hydroxyecdysone, 25-Deoxyecdysterone, 2beta,3beta,14alpha,20R,22R-Pentahydroxy-5beta-cholest-7-en-6-one. Purity: ≥95% (HPLC). Solubility: Soluble in acetic acid, 100% ethanol, methanol or DMSO; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8895