Learn More
Eurofins Discoverx Ponatinib, 10 mg. (92-1315)
Supplier:  Eurofins Discoverx 921315
Encompass
Ponatinib (or Iclusig) is typically used for the treatment of chronic myeloid leukemia and Philadelphia chromosome–positive (Ph+) acute lymphoblastic leukemia. It is intended to be effective against T315I mutated CML. Ponatinib is an inhibitor of multiple tyrosine kinases, including RIPK1, 2, and 3. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9828