Learn More
Medchemexpress LLC HY-12047 50mg Medchemexpress, Ponatinib CAS:943319-70-8 Purity:>98%
SDP
Catalog No. 502028844
Encompass_Preferred
Medchemexpress, HY-12047 50mg Ponatinib CAS:943319-70-8 Ponatinib (AP24534) is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM, 1.1 nM, 1.5 nM, 2.2 nM, and 5.4 nM for Abl, PDGFRα, VEGFR2, FGFR1, and Src, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8844 Supplier Medchemexpress LLC Supplier No. HY1204750MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-12047 50mg Ponatinib CAS:943319-70-8 Ponatinib (AP24534) is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM, 1.1 nM, 1.5 nM, 2.2 nM, and 5.4 nM for Abl, PDGFRα, VEGFR2, FGFR1, and Src, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.