Learn More
Apexbio Technology LLC Ponatinib (AP24534) 943319-70-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A54675.1
Encompass_Preferred
Ponatinib (AP24534 CAS 943319-70-8) is a second-generation small molecule inhibitor targeting the BCR-ABL fusion kinase The BCR-ABL fusion arising from a t(9 22) chromosomal translocation encodes oncogenic tyrosine kinase variants implicated in chronic myeloid leukemia (CML) acute lymphoblastic leukemia (ALL) and acute myeloid leukemia (AML) Ponatinib inhibits both wild-type BCR-ABL (IC50 0 5 nM) and resistant mutants notably the T315I mutant (IC50 11 nM) Additionally it potently inhibits clinically relevant kinases such as RET FLT3 KIT PDGFR / and FGFR1 Ponatinib is utilized in cancer biology research pertaining to kinase-driven malignancies
Catalog No. 50-000-62336