Learn More
Apexbio Technology LLC Ponatinib (AP24534), 25mg. Cas: 943319-70-8 MFCD: MFCD17215203. pan-BCR-ABL inhibitor,multi-kinase inhibitor
SDP
Supplier:  Apexbio Technology LLC A546725
Encompass_Preferred
BCR-ABL fusion gene forms when the ABL gene from chromosome 9 joins to the BCR gene on chromosome 22. BCR-ABL is translated into a constitutively active tyrosine kinase, which is oncogenic. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4953