Learn More
Apexbio Technology LLC Ponatinib (AP24534), 943319-70-8, MFCD17215203, 5 mg
SDP
Supplier:  Apexbio Technology LLC A54675
Encompass_Preferred
BCR-ABL fusion gene forms when the ABL gene from chromosome 9 joins to the BCR gene on chromosome 22. BCR-ABL is translated into a constitutively active tyrosine kinase, which is oncogenic. Depending on the fusion location, multiple protein variants are formed with molecular weight ranging from 185 to 210 kDa. BCR-ABL activates JAK/STAT pathway and MAPK signaling. [3] This gene is found in most patients with chronic myelogenous leukemia (CML), and in some patients with acute lymphoblastic leukemia (ALL) or acute myelogenous leukemia (AML). Ponatinib is the second-generation pan inhibitor of BCR-Abl kinases, which is also effective against the mutant form of BCR-Abl (T315I). IC50 for WT and mutant form are 0.5 and 11 nM. [4] Ponatinib also inhibits several other clinically relevant kinases (RET, FLT3, KIT, PDGFRα, PDGFRβ, and FGFR1) in vitro, with IC50s of 5, 25, 100, 5, 9, and 23) in Ba/F3 cells lines. Other sizes are available. Please inqury us for quote.
Catalog No. 50-101-3061