Learn More
Medchemexpress LLC Ponatinib-d8 (AP24534-d8) | 1562993-37-6 | 99.9% | 1 MG
SDP
Catalog No. 5000234374
Encompass_Preferred
Ponatinib-d8 is a deuterium-labeled analogue of ponatinib used as a stable isotope internal standard for quantitative mass spectrometry and pharmacokinetic assays. It provides accurate normalization for LC-MS/MS workflows and supports method validation and PK sample analysis.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High chemical and isotopic purity for reliable quantitation.
  • Suitable for bioanalytical method validation and PK studies.
  • Offered in small, convenient quantities for analytical use.
Catalog No. 50-002-34374 Supplier Medchemexpress LLC Supplier No. HY12047S1MG
Ponatinib-d8 is a deuterium-labeled analogue of ponatinib used as a stable isotope internal standard for quantitative mass spectrometry and pharmacokinetic assays. It provides accurate normalization for LC-MS/MS workflows and supports method validation and PK sample analysis.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High chemical and isotopic purity for reliable quantitation.
  • Suitable for bioanalytical method validation and PK studies.
  • Offered in small, convenient quantities for analytical use.