Learn More
TARGETMOL CHEMICALS INC PONATINIB HYDROCHLORIDE 10MG
SDP
Catalog No. 502580849
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Ponatinib Hydrochloride (AP-24534 Hydrochloride) is a hydrochloride of ponatinib. Ponatinib is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM 1.1 nM 1.5 nM 2.2 nM and 5.4 nM for Abl PDGFR(alpha) VEGFR2 FGFR1 and Src respectively. purity: 100%
Catalog No. 50-258-0849 Supplier TARGETMOL CHEMICALS INC Supplier No. T2372L10MG
Also available in 25 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Ponatinib Hydrochloride (AP-24534 Hydrochloride) is a hydrochloride of ponatinib. Ponatinib is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM 1.1 nM 1.5 nM 2.2 nM and 5.4 nM for Abl PDGFR(alpha) VEGFR2 FGFR1 and Src respectively. purity: 100%