Learn More
Medchemexpress LLC Ponatinib (hydrochloride) | 1114544-31-8 | 99.8% | 569.02 | 10 MG
SDP
Catalog No. 5000344558
Encompass_Preferred
Ponatinib hydrochloride is a hydrochloride of ponatinib, an orally active multi-targeted kinase inhibitor. It targets Abl, PDGFRα, VEGFR2, FGFR1, and Src with various IC50s.
  • Reduces the viability, migration, and function of human endothelial cells.
  • Decreased cell viability in HUVEC cells in a dose-dependent manner.
  • Increased the percentage of annexin/PI labeled cells in HUVEC cells.
  • Reduced VEGF-induced migration in HUVEC cells.
  • Protects mice from lethal influenza infection by inhibiting cytokine storm.
  • Treated mice had higher survival rates and reduced the infiltration of neutrophils.
Catalog No. 50-003-44558 Supplier Medchemexpress LLC Supplier No. HY10876610MM/1ML
Ponatinib hydrochloride is a hydrochloride of ponatinib, an orally active multi-targeted kinase inhibitor. It targets Abl, PDGFRα, VEGFR2, FGFR1, and Src with various IC50s.
  • Reduces the viability, migration, and function of human endothelial cells.
  • Decreased cell viability in HUVEC cells in a dose-dependent manner.
  • Increased the percentage of annexin/PI labeled cells in HUVEC cells.
  • Reduced VEGF-induced migration in HUVEC cells.
  • Protects mice from lethal influenza infection by inhibiting cytokine storm.
  • Treated mice had higher survival rates and reduced the infiltration of neutrophils.