Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ponatinib hydrochloride | 1114544-31-8 | 99.8% | 569.02 | 25 MG
SDP

Catalog No. 5000347270
Encompass_Preferred

Ponatinib hydrochloride, also known as AP24534 hydrochloride, is an orally active multi-targeted kinase inhibitor. It inhibits various kinases with IC50s including 0.37 nM for Abl, 1.1 nM for PDGFRα, 1.5 nM for VEGFR2, 2.2 nM for FGFR1, and 5.4 nM for Src.

  • Purity of 99.76%
  • CAS number: 1114544-31-8
  • Molecular weight: 569.02
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 25 mg/mL
  • Targets Bcr-Abl, FGFR, PDGFR, VEGFR, Src, and Autophagy pathways
  • Store at 4°C, sealed, away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-47270 Supplier Medchemexpress LLC Supplier No. 25MGHY108766
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ponatinib hydrochloride, also known as AP24534 hydrochloride, is an orally active multi-targeted kinase inhibitor. It inhibits various kinases with IC50s including 0.37 nM for Abl, 1.1 nM for PDGFRα, 1.5 nM for VEGFR2, 2.2 nM for FGFR1, and 5.4 nM for Src.

  • Purity of 99.76%
  • CAS number: 1114544-31-8
  • Molecular weight: 569.02
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 25 mg/mL
  • Targets Bcr-Abl, FGFR, PDGFR, VEGFR, Src, and Autophagy pathways
  • Store at 4°C, sealed, away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.