Learn More
Medchemexpress LLC Ponatinib (hydrochloride) | 1114544-31-8 | MFCD28656903 | 98.0% | 569.02 g·mol⁻¹ | C29H28ClF3N6O | 5 MG

Supplier: Medchemexpress LLC HY1087665MG
Ponatinib hydrochloride is the hydrochloride salt of ponatinib, supplied as an analytical research standard intended for laboratory and assay development use. The material is provided as a solid reference standard with accompanying batch documentation for traceability.
- Research-grade hydrochloride salt suitable for analytical applications.
- Purity 98.0% as reported in the certificate of analysis.
- Molecular formula C29H28ClF3N6O; molecular weight 569.02 g·mol⁻¹.
- Supplied as a 5 mg solid vial for small-scale assays.
- Provided with SMILES and batch documentation for structural reference.
- Intended for research use only, not for human or veterinary applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.