Learn More
Medchemexpress LLC Ponatinib hydrochloride (AP24534 hydrochloride) | 1114544-31-8 | 99.76% | 569.02 | 500 MG
SDP
Catalog No. 5000345096
Encompass_Preferred
Ponatinib hydrochloride (AP24534 hydrochloride) is a hydrochloride salt of ponatinib. It is an orally active multi-targeted kinase inhibitor with IC50 values of 0.37 nM for Abl, 1.1 nM for PDGFRα, 1.5 nM for VEGFR2, 2.2 nM for FGFR1, and 5.4 nM for Src.
  • Orally active multi-targeted kinase inhibitor.
  • Inhibits Abl with an IC50 of 0.37 nM.
  • Targets PDGFRα with an IC50 of 1.1 nM.
  • Inhibits VEGFR2 with an IC50 of 1.5 nM.
  • Targets FGFR1 with an IC50 of 2.2 nM.
  • Inhibits Src with an IC50 of 5.4 nM.
  • Reduced viability, migration, and function of human endothelial cells in vitro.
  • Protects mice from lethal influenza infection by inhibiting cytokine storm in vivo.
Catalog No. 50-003-45096 Supplier Medchemexpress LLC Supplier No. HY108766100MG
Ponatinib hydrochloride (AP24534 hydrochloride) is a hydrochloride salt of ponatinib. It is an orally active multi-targeted kinase inhibitor with IC50 values of 0.37 nM for Abl, 1.1 nM for PDGFRα, 1.5 nM for VEGFR2, 2.2 nM for FGFR1, and 5.4 nM for Src.
  • Orally active multi-targeted kinase inhibitor.
  • Inhibits Abl with an IC50 of 0.37 nM.
  • Targets PDGFRα with an IC50 of 1.1 nM.
  • Inhibits VEGFR2 with an IC50 of 1.5 nM.
  • Targets FGFR1 with an IC50 of 2.2 nM.
  • Inhibits Src with an IC50 of 5.4 nM.
  • Reduced viability, migration, and function of human endothelial cells in vitro.
  • Protects mice from lethal influenza infection by inhibiting cytokine storm in vivo.