Learn More
Medchemexpress LLC Ponatinib hydrochloride (AP24534 hydrochloride) | 1114544-31-8 | 99.8% | 569.02 | 100 U
SDP
Catalog No. 5000345437
Encompass_Preferred
Ponatinib hydrochloride is a hydrochloride of ponatinib. Ponatinib is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM, 1.1 nM, 1.5 nM, 2.2 nM, and 5.4 nM for Abl, PDGFRα, VEGFR2, FGFR1, and Src, respectively. It is for research use only and not sold to patients.
  • Orally active multi-targeted kinase inhibitor
  • IC50s: 0.37 nM for Abl, 1.1 nM for PDGFRα, 1.5 nM for VEGFR2, 2.2 nM for FGFR1, and 5.4 nM for Src
  • Reduces viability, migration, and function of human endothelial cells in vitro
  • Protects mice from lethal influenza infection by inhibiting cytokine storm in vivo
Catalog No. 50-003-45437 Supplier Medchemexpress LLC Supplier No. HY10876650MG
Ponatinib hydrochloride is a hydrochloride of ponatinib. Ponatinib is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM, 1.1 nM, 1.5 nM, 2.2 nM, and 5.4 nM for Abl, PDGFRα, VEGFR2, FGFR1, and Src, respectively. It is for research use only and not sold to patients.
  • Orally active multi-targeted kinase inhibitor
  • IC50s: 0.37 nM for Abl, 1.1 nM for PDGFRα, 1.5 nM for VEGFR2, 2.2 nM for FGFR1, and 5.4 nM for Src
  • Reduces viability, migration, and function of human endothelial cells in vitro
  • Protects mice from lethal influenza infection by inhibiting cytokine storm in vivo