Learn More
Medchemexpress LLC Ponceau 4R | 2611-82-7 | 85.0% | 50 G
SDP
Catalog No. 5000410678
Encompass_Preferred
Ponceau 4R (85%) is a synthetic colorant, a strawberry red azo dye that can be used as a food coloring in various food products. It is typically synthesized from aromatic hydrocarbons and exhibits stability to light, heat, and acid, though it will fade when exposed to ascorbic acid.
  • Synthetic colorant
  • Strawberry red azo dye
  • Can be used as a food coloring
  • Stable to light, heat, and acid
  • Fades in the presence of ascorbic acid
  • Appears as a pink to red solid
  • Soluble in water at 100 mg/mL
Catalog No. 50-004-10678 Supplier Medchemexpress LLC Supplier No. HYD0193A50G
Ponceau 4R (85%) is a synthetic colorant, a strawberry red azo dye that can be used as a food coloring in various food products. It is typically synthesized from aromatic hydrocarbons and exhibits stability to light, heat, and acid, though it will fade when exposed to ascorbic acid.
  • Synthetic colorant
  • Strawberry red azo dye
  • Can be used as a food coloring
  • Stable to light, heat, and acid
  • Fades in the presence of ascorbic acid
  • Appears as a pink to red solid
  • Soluble in water at 100 mg/mL