Learn More
Electron Microscopy Sciences Ponceau Acid Fuchsin Solution 250 ML
SDP
Catalog No. 5031791
Encompass_Preferred
Reference: For the staining of fibrin and platelets. Carstairs (1965).
Catalog No. 50-317-91 Supplier Electron Microscopy Sciences Supplier No. 2638104
Reference: For the staining of fibrin and platelets. Carstairs (1965).