Learn More
Abcam Ponceau S, Protein dye, 25G
Catalog No. NC1768354
Encompass_Preferred
MW 760.61 g/mol. Protein dye for the rapid, reversible staining of protein bands on nitrocellulose, PVDF and cellulose acetate membranes. Staining is reversible allowing further immunological detection. Negative stain which binds to the positively charged amino groups of the protein. Also binds non-covalently to non-polar regions in the protein. (Note: Ponceau S is not suitable for use with nylon membranes.).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1768354 Supplier Abcam Supplier No. AB14631325G
MW 760.61 g/mol. Protein dye for the rapid, reversible staining of protein bands on nitrocellulose, PVDF and cellulose acetate membranes. Staining is reversible allowing further immunological detection. Negative stain which binds to the positively charged amino groups of the protein. Also binds non-covalently to non-polar regions in the protein. (Note: Ponceau S is not suitable for use with nylon membranes.).
The product is subject to the following: Abcam Restricted Use Statement