Learn More
Crescent Chemical Co Inc PONCEAU S
SDP
Catalog No. 50248054
Encompass_Preferred
Ponceau S For reversible protein staining on membranes and for microscopy. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 33429.02 CAS 6226-79-5
Catalog No. 50-248-054 Supplier Crescent Chemical Co Inc Supplier No. SE33429.02
Ponceau S For reversible protein staining on membranes and for microscopy. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 33429.02 CAS 6226-79-5