Learn More
Crescent Chemical Co Inc PONCEAU S
Supplier: Crescent Chemical Co Inc SE33429.01
Ponceau S For reversible protein staining on membranes and for microscopy. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 33429.01 CAS 6226-79-5