Learn More
Biotang Inc PONCEAU S, 500 ML, ST-180, Ponceau S solution 0.1% (w/v) PONCEAU S in 5% acetic acid. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50152421 Shop All BIOTANG Inc Products
Encompass_Preferred
PONCEAU S, 500 ML, ST-180, Ponceau S solution 0.1% (w/v) PONCEAU S in 5% acetic acid. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-421 Supplier Biotang Inc Supplier No. BTBB967
PONCEAU S, 500 ML, ST-180, Ponceau S solution 0.1% (w/v) PONCEAU S in 5% acetic acid. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped at room temperature. Store at room temperature.