Learn More
Medchemexpress LLC Ponceau S (acid red 112) | 6226-79-5 | MFCD00003892 | 98.0% | 760.57 g/mol | C22H12N4Na4O13S4 | 100G
SDP
Catalog No. 5000239503
Encompass_Preferred
Ponceau S is a reversible, non-specific protein stain supplied as a powder for preparation of an acidic aqueous staining solution. It provides rapid visualization of protein bands on nitrocellulose and PVDF membranes and can be removed to allow downstream immunodetection workflows without interfering with antibody-antigen binding.
  • Rapid, reversible visualization of protein bands.
  • Compatible with antibody-antigen binding and downstream immunodetection.
  • Easy to prepare as an acidic aqueous staining solution.
  • Suitable for nitrocellulose and PVDF membranes.
  • Available in multiple package sizes for laboratory use.
Catalog No. 50-002-39503 Supplier Medchemexpress LLC Supplier No. HY12489A100G
Ponceau S is a reversible, non-specific protein stain supplied as a powder for preparation of an acidic aqueous staining solution. It provides rapid visualization of protein bands on nitrocellulose and PVDF membranes and can be removed to allow downstream immunodetection workflows without interfering with antibody-antigen binding.
  • Rapid, reversible visualization of protein bands.
  • Compatible with antibody-antigen binding and downstream immunodetection.
  • Easy to prepare as an acidic aqueous staining solution.
  • Suitable for nitrocellulose and PVDF membranes.
  • Available in multiple package sizes for laboratory use.