Learn More
Medchemexpress LLC Ponceau S, sodium salt | 6226-79-5 | MFCD00003892 | 75.0% | 760.57 g/mol | C22H12N4Na4O13S4 | 25G
SDP
Catalog No. 5000236530
Encompass_Preferred
Ponceau S (sodium salt) is a water-soluble azo dye used for rapid, reversible staining of proteins on nitrocellulose, PVDF, and cellulose acetate membranes. Supplied as a powder for solution preparation, it is commonly used to visualize protein bands during membrane transfer checks and prior to immunodetection workflows, and can be removed without affecting downstream analyses.
  • Provides rapid, reversible staining of protein bands on membranes.
  • Compatible with nitrocellulose, PVDF, and cellulose acetate membranes.
  • Water-soluble dye formulated for acidic staining solutions.
  • Supplied as a powder for on-demand solution preparation and storage.
  • Typical dye content 75% for routine visualization.
  • Removes easily to permit subsequent immunodetection steps.
Catalog No. 50-002-36530 Supplier Medchemexpress LLC Supplier No. HY12489A25G
Ponceau S (sodium salt) is a water-soluble azo dye used for rapid, reversible staining of proteins on nitrocellulose, PVDF, and cellulose acetate membranes. Supplied as a powder for solution preparation, it is commonly used to visualize protein bands during membrane transfer checks and prior to immunodetection workflows, and can be removed without affecting downstream analyses.
  • Provides rapid, reversible staining of protein bands on membranes.
  • Compatible with nitrocellulose, PVDF, and cellulose acetate membranes.
  • Water-soluble dye formulated for acidic staining solutions.
  • Supplied as a powder for on-demand solution preparation and storage.
  • Typical dye content 75% for routine visualization.
  • Removes easily to permit subsequent immunodetection steps.