Learn More
Abcam Ponceau S Solution, 1000ML
Catalog No. NC1914944
Encompass_Preferred
Ponceau S Solution (ab270042) is a ready-to-use reversible stain which allows you to check the efficiency of electrophoretic transfer of SDS-denatured proteins to nitrocellulose and PVDF membranes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1914944 Supplier Abcam Supplier No. AB2700421000ML
Ponceau S Solution (ab270042) is a ready-to-use reversible stain which allows you to check the efficiency of electrophoretic transfer of SDS-denatured proteins to nitrocellulose and PVDF membranes.
The product is subject to the following: Abcam Restricted Use Statement