Learn More
Sigma Aldrich Fine Chemicals Biosciences Ponceau S solution, 6226-79-5, MFCD00003892, 1L
Catalog No. 501657080
Encompass
Empirical Formula (Hill Notation) C22H12N4Na4O13S4, Molecular Weight 760.57, BioReagent, suitable for electrophoresis, 0.1 % (w/v) in 5% acetic acid
Catalog No. 50-165-7080 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P71701L
Empirical Formula (Hill Notation) C22H12N4Na4O13S4, Molecular Weight 760.57, BioReagent, suitable for electrophoresis, 0.1 % (w/v) in 5% acetic acid