Learn More
Crescent Chemical Co Inc Ponceau S Solution for Electrophoresis (0.2%), 500mL
SDP
Catalog No. 50248052
Encompass_Preferred
Ponceau S Solution for Electrophoresis (0.2%), 500mL, In 3 % TCA. For reversible protein staining on membranes. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-052 Supplier Crescent Chemical Co Inc Supplier No. SE33427.01
Ponceau S Solution for Electrophoresis (0.2%), 500mL, In 3 % TCA. For reversible protein staining on membranes. Storage Conditions: +15 °C TO +30 °C