Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ponceau S Staining Solution

Catalog No. A40000278 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
3 x 1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40000278 1000 mL
A40000279 500 mL
A40000278X3 3 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A40000278 Supplier Thermo Scientific™ Supplier No. A40000278
  • $141.00
    $123.65 / Each of 1

    Save $17.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.

Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Form Liquid
Formulation 0.1% (w/v) in 5% acetic acid
For Use With (Application) Western Blotting
Quantity 1000 mL
Product Type Ponceau S Staining Solution
What is the best way to image Ponceau staining with an iBright Imaging System?

The recommended imaging method depends on the iBright imager model being used. For iBright FL1500 and CL1500 Imaging Systems, we recommend imaging Ponceau-stained samples with the Ponceau application that can be found in Universal mode. For the iBright CL750 Imaging System, we recommend imaging Ponceau-stained samples in Protein gel mode.

We always recommend imaging with the orange iBright System Contrast Tray (Cat. No. A56597), and if needed, adjust the focus or zoom to get bands as sharp as possible. Better quality pictures are obtained when the membrane is wet.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.