Learn More
Research Products International Corp Ponceau S Staining Solution, 500 Milliliters
SDP
Catalog No. 501368540
Encompass_Preferred
Composition:
Water: ≥ 94.9%
Acetic Acid: 5.0%
Ponceau S: 0.1%
Catalog No. 50-136-8540 Supplier Research Products International Corp Supplier No. P56200500.0
Composition:
Water: ≥ 94.9%
Acetic Acid: 5.0%
Ponceau S: 0.1%