Learn More
Cayman Chemical Ponceau S (sodium salt), CAS: 6226-79-5, 50 g
SDP
Catalog No. 502953896
Encompass_Preferred
A stain that permits the rapid, reversible detection of proteins immobilized on various matrices; used to locate proteins or evaluate efficiency of transfer, or an alternative to actin probing as a loading control; also used as a fiducial marker in mass spectrometric imaging
Catalog No. 50-295-3896 Supplier Cayman Chemical Supplier No. 1433050 G
A stain that permits the rapid, reversible detection of proteins immobilized on various matrices; used to locate proteins or evaluate efficiency of transfer, or an alternative to actin probing as a loading control; also used as a fiducial marker in mass spectrometric imaging