Learn More
Medchemexpress LLC Poncirin | 14941-08-3 | 5 MG
SDP
Catalog No. 5000418736
Encompass_Preferred
Poncirin is a natural product isolated from Poncirus trifoliata with anti-inflammatory activities. It significantly reduces mechanical hyperalgesia and allodynia in Complete Freund's Adjuvant (CFA)-induced inflammatory pain models, and has been shown to reduce pain behavior in acetic acid-induced visceral pain and formalin-induced tonic pain models.
  • Isolated from Poncirus trifoliata
  • Possesses anti-inflammatory activities
  • Reduces mechanical hyperalgesia and allodynia
  • Effective in CFA-induced inflammatory pain models
  • Reduces pain in acetic acid-induced visceral pain models
  • Reduces pain in formalin-induced tonic pain models
Catalog No. 50-004-18736 Supplier Medchemexpress LLC Supplier No. HYN22585MG
Poncirin is a natural product isolated from Poncirus trifoliata with anti-inflammatory activities. It significantly reduces mechanical hyperalgesia and allodynia in Complete Freund's Adjuvant (CFA)-induced inflammatory pain models, and has been shown to reduce pain behavior in acetic acid-induced visceral pain and formalin-induced tonic pain models.
  • Isolated from Poncirus trifoliata
  • Possesses anti-inflammatory activities
  • Reduces mechanical hyperalgesia and allodynia
  • Effective in CFA-induced inflammatory pain models
  • Reduces pain in acetic acid-induced visceral pain models
  • Reduces pain in formalin-induced tonic pain models