Learn More
Medchemexpress LLC Poneratoxin acetate | 2756.35 (free base) | 10 MG
SDP
Catalog No. 5000425502
Encompass_Preferred
Poneratoxin acetate is the acetate salt form of Poneratoxin (HY-P10234). It acts as a modulator for voltage-gated sodium channels (NaV, with EC50 for NaV1.6 and NaV1.7 being 97 nM and 2.3 μM), decreasing the voltage threshold for activation and inhibiting channel inactivation. This compound enhances the excitability of neurons and contributes to the transmission of pain signals.
  • Modulator for voltage-gated sodium channels
  • Lowers the voltage threshold for activation
  • Inhibits the inactivation of channels
  • Enhances the excitability of neurons
  • Contributes to the transmission of pain signals
  • For powder, store at -80°C for 2 years or -20°C for 1 year
  • For solution, store at -80°C for 6 months or -20°C for 1 month
  • Store sealed and away from moisture
Catalog No. 50-004-25502 Supplier Medchemexpress LLC Supplier No. HYP10234A10MG
Poneratoxin acetate is the acetate salt form of Poneratoxin (HY-P10234). It acts as a modulator for voltage-gated sodium channels (NaV, with EC50 for NaV1.6 and NaV1.7 being 97 nM and 2.3 μM), decreasing the voltage threshold for activation and inhibiting channel inactivation. This compound enhances the excitability of neurons and contributes to the transmission of pain signals.
  • Modulator for voltage-gated sodium channels
  • Lowers the voltage threshold for activation
  • Inhibits the inactivation of channels
  • Enhances the excitability of neurons
  • Contributes to the transmission of pain signals
  • For powder, store at -80°C for 2 years or -20°C for 1 year
  • For solution, store at -80°C for 6 months or -20°C for 1 month
  • Store sealed and away from moisture