Learn More
Selleck Chemical LLC Ponesimod 5mg 854107-55-4 ACT-128800
SDP
Catalog No. 501365838
Encompass_Preferred
Ponesimod (ACT-128800) is an orally active, selective sphingosine-1-phosphate receptor 1 (S1P1) immunomodulator with EC50 of 5.7 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5838 Supplier Selleck Chemical LLC Supplier No. S82415MG
Ponesimod (ACT-128800) is an orally active, selective sphingosine-1-phosphate receptor 1 (S1P1) immunomodulator with EC50 of 5.7 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.