Learn More
Medchemexpress LLC Ponesimod-d7 | 00-00-0 | 96.5% | 468.02 g/mol | C23H18D7ClN2O4S | 50 MG
SDP
Catalog No. 5000240064
Encompass_Preferred
Ponesimod-d7 is a deuterium-labeled isotopologue of ponesimod supplied as a 50 mg reference standard for analytical and research use. It is intended for use as a stable-isotope internal standard in mass spectrometry, tracer experiments, and pharmacokinetic studies; store and handle according to supplier recommendations.
  • Deuterium-labeled isotopologue for use as a stable-isotope reference.
  • Suitable as an internal standard for LC-MS/MS quantitation.
  • High isotopic enrichment to support accurate tracer studies.
  • Supplied as a 50 mg powder with recommended storage at -20°C.
  • Useful for pharmacokinetic and metabolite identification studies.
Catalog No. 50-002-40064 Supplier Medchemexpress LLC Supplier No. HY10569S150MG
Ponesimod-d7 is a deuterium-labeled isotopologue of ponesimod supplied as a 50 mg reference standard for analytical and research use. It is intended for use as a stable-isotope internal standard in mass spectrometry, tracer experiments, and pharmacokinetic studies; store and handle according to supplier recommendations.
  • Deuterium-labeled isotopologue for use as a stable-isotope reference.
  • Suitable as an internal standard for LC-MS/MS quantitation.
  • High isotopic enrichment to support accurate tracer studies.
  • Supplied as a 50 mg powder with recommended storage at -20°C.
  • Useful for pharmacokinetic and metabolite identification studies.