Learn More
Medchemexpress LLC Ponesimod-d7 | 00-00-0 | MFCD18207776 | 468.02 g·mol⁻1 | C23H18D7ClN2O4S | 5 MG
SDP
Catalog No. 5000240516
Encompass_Preferred
Ponesimod-d7 is the deuterium-labeled analogue of ponesimod provided for research use as an isotopic tracer. It retains the parent compound's pharmacological profile while introducing a mass shift that enables unambiguous detection in mass-spectrometry assays. Typical properties include molecular formula C23H18D7ClN2O4S and molecular weight 468.02 g·mol⁻1.
  • Deuterium-labeled analogue suitable for tracer studies.
  • Enables mass-shift detection in LC-MS assays.
  • Useful for metabolic and pharmacokinetic investigations.
  • Provided as a stable powder for analytical workflows.
  • Storage recommendations: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
Catalog No. 50-002-40516 Supplier Medchemexpress LLC Supplier No. HY10569S15MG
Ponesimod-d7 is the deuterium-labeled analogue of ponesimod provided for research use as an isotopic tracer. It retains the parent compound's pharmacological profile while introducing a mass shift that enables unambiguous detection in mass-spectrometry assays. Typical properties include molecular formula C23H18D7ClN2O4S and molecular weight 468.02 g·mol⁻1.
  • Deuterium-labeled analogue suitable for tracer studies.
  • Enables mass-shift detection in LC-MS assays.
  • Useful for metabolic and pharmacokinetic investigations.
  • Provided as a stable powder for analytical workflows.
  • Storage recommendations: powder at -20°C or 4°C; in solvent at -80°C or -20°C.