Learn More
Medchemexpress LLC Ponesimod-d7 | 00-00-0 | MFCD18207776 | 96.5% | 468.02 | C23H18D7ClN2O4S | 10 MG
SDP
Catalog No. 5000240189
Encompass_Preferred
Ponesimod-d7 is the deuterium-labeled analogue of ponesimod provided for research and analytical use. This isotopically labeled compound acts as a selective S1P1 receptor agonist and is intended primarily for use as an internal standard, tracer, or reference material in pharmacokinetic and metabolic studies. It is supplied with high isotopic enrichment and high chemical purity suitable for analytical workflows.
  • Deuterium-labeled analogue for tracer and internal standard applications.
  • Selective S1P1 receptor activity suitable for biological assays.
  • High isotopic enrichment (d7) for clear mass spectrometry distinction.
  • High chemical purity appropriate for analytical and research use.
  • Supplied in small, convenient quantities for method development and analysis.
Catalog No. 50-002-40189 Supplier Medchemexpress LLC Supplier No. HY10569S110MG
Ponesimod-d7 is the deuterium-labeled analogue of ponesimod provided for research and analytical use. This isotopically labeled compound acts as a selective S1P1 receptor agonist and is intended primarily for use as an internal standard, tracer, or reference material in pharmacokinetic and metabolic studies. It is supplied with high isotopic enrichment and high chemical purity suitable for analytical workflows.
  • Deuterium-labeled analogue for tracer and internal standard applications.
  • Selective S1P1 receptor activity suitable for biological assays.
  • High isotopic enrichment (d7) for clear mass spectrometry distinction.
  • High chemical purity appropriate for analytical and research use.
  • Supplied in small, convenient quantities for method development and analysis.