Learn More
Medchemexpress LLC Ponesimod-d7 100ug | 100UG
SDP
Catalog No. 5000241547
Encompass_Preferred
Ponesimod-d7 (ACT-128800-d7) is the deuterium-labeled Ponesimod (HY-10569) Ponesimod-d7 (ACT-128800) is a potent selective and orally active agonist of S1P1 with an IC50 of 6 nM in a radioligand binding assay Ponesimod-d7 activates S1P1-mediated signal transduction with high potency (EC50 5 7 nM) Ponesimod-d7 can protect against lymphocyte-mediated tissue inflammation[1 [2 [3
Catalog No. 50-002-41547 Supplier Medchemexpress LLC Supplier No. HY10569S1100UG
Ponesimod-d7 (ACT-128800-d7) is the deuterium-labeled Ponesimod (HY-10569) Ponesimod-d7 (ACT-128800) is a potent selective and orally active agonist of S1P1 with an IC50 of 6 nM in a radioligand binding assay Ponesimod-d7 activates S1P1-mediated signal transduction with high potency (EC50 5 7 nM) Ponesimod-d7 can protect against lymphocyte-mediated tissue inflammation[1 [2 [3