Learn More
Abcam Ponesimod, sphingosine-1-phophate receptor agonist, 1MG
Catalog No. 502672633
Encompass_Preferred
MW 461 Da. Ponesimod is a potent agonist of sphingosine-1-phophate receptor. (S1P₁/EDG-1; IC₅₀s = 6, >10,000, 2,068, 1,956, and 142 nM for S1P₁-S1P₅, respectively, in a radioligand binding assay). Selectively activates S1P₁ in a GTPγS assay (EC₅₀s = 5.7, >10,000, 105, 1,108, and 59.1 nM, for S1P₁-S1P₅, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2633 Supplier Abcam Supplier No. AB2315091MG
MW 461 Da. Ponesimod is a potent agonist of sphingosine-1-phophate receptor. (S1P₁/EDG-1; IC₅₀s = 6, >10,000, 2,068, 1,956, and 142 nM for S1P₁-S1P₅, respectively, in a radioligand binding assay). Selectively activates S1P₁ in a GTPγS assay (EC₅₀s = 5.7, >10,000, 105, 1,108, and 59.1 nM, for S1P₁-S1P₅, respectively).
The product is subject to the following: Abcam Restricted Use Statement