Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pontacyl Carmine 2B 93.0+%, TCI America™
SDP

Catalog No. P059525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P059525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P059525G Supplier TCI America Supplier No. P059525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6625-46-3
Molecular Formula C19H15N3Na2O9S2
Molecular Weight (g/mol) 539.441
MDL Number MFCD00059131
InChI Key JIAWPRMYSNTTFB-UHFFFAOYSA-L
PubChem CID 73555245
IUPAC Name disodium;5-acetamido-3-[(2-methoxyphenyl)hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
SMILES CC(=O)NC1=C2C(=CC(=C1)S(=O)(=O)[O-])C=C(C(=NNC3=CC=CC=C3OC)C2=O)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Red
Quantity 25 g
Formula Weight 539.44
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pontacyl Carmine 2B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.